Invitrogen
Manufacturer Code:PA540771
Catalog # PIPA540771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MED27 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cofactor required for Sp1 transcriptional activation subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8, 34kDa; CRSP complex subunit 8; Mediator complex subunit 27; Mediator complex subunit 27 CRAP34 CRSP34 CRSP8 MGC11274 TRAP37; Mediator of RNA polymerase II transcription subunit 27; P37 TRAP/SMCC/PC2 subunit; transcriptional co-activator CRSP8 homolog; Transcriptional coactivator CRSP34
Gene Aliases: 1500015J03Rik; 2310042P07Rik; AA682045; CRAP34; CRSP34; CRSP8; D2Ertd434e; MED27; MED3; RGD1564993; TRAP37
UniProt ID: (Bovine) Q2TBN7, (Human) Q6P2C8, (Mouse) Q9DB40
Entrez Gene ID: (Dog) 612800, (Horse) 100069513, (Bovine) 525389, (Rabbit) 100357013, (Human) 9442, (Guinea pig) 100725304, (Rat) 296612, (Mouse) 68975
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.