Abnova
Manufacturer Code:H00009442K
Catalog # 89-209-984
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MED27 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MED27 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Cofactor required for Sp1 transcriptional activation subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8, 34kDa; CRAP34 CRSP34 CRSP8 MGC11274 TRAP37; CRSP complex subunit 8; Mediator complex subunit 27; Mediator of RNA polymerase II transcription subunit 27; P37 TRAP/SMCC/PC2 subunit; Transcriptional coactivator CRSP34
Gene Aliases: CRAP34; CRSP34; CRSP8; MED27; MED3; TRAP37
UniProt ID: (Human) Q6P2C8
Entrez Gene ID: (Human) 9442
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.