OriGene
Manufacturer Code:CF810428
Catalog # 50-167-0065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D1 |
Immunogen | Full length human recombit protein of human MED19 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DT2P1G7 LCMR1 MED19AS; Lung cancer metastasis-related protein 1; Mediator complex subunit 19; Mediator of RNA polymerase II transcription subunit 19; mediator of RNA polymerase II transcription, subunit 19 homolog; putative mediator subunit MED19AS protein
Gene Aliases: DT2P1G7; LCMR1; MED19; MED19AS
UniProt ID: (Human) A0JLT2
Entrez Gene ID: (Human) 219541
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.