Novus Biologicals
Manufacturer Code:NBP245780
Catalog # NBP245780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C11 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 393-679 of human MED15(NP_056973) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activator-recruited cofactor 105 kDa component; activator-recruited cofactor, 105-kD; ARC105; ARC105Arc105 CAG7A CTG repeat protein 7a CTG7A FLJ42282 FLJ42935 mediator complex subunit 15PC2 glutamine/Q-rich-associated protein mediator of RNA polymerase II transcription subunit 15 PC2 (positive cofactor 2 multiprotein complex) glutamine/Q-rich-associatedprotein PC2-glutamine-rich-associated protein PCQAPDKFZp686A2214 Positive cofactor 2 glutamine/Q-rich-associated protein positive cofactor 2 glutamine/Q-rich-associated protein TIG1DKFZp762B1216 TIG-1TPA-inducible gene 1 protein TNRC7 TPA inducible gene-1 TPA inducible protein trinucleotide repeat containing 7105-kD Trinucleotide repeat-containing gene 7 protein; CTG repeat protein 7a; Mediator complex subunit 15; Mediator of RNA polymerase II transcription subunit 15; PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associated protein; PC2 glutamine/Q-rich-associated protein; PC2-glutamine-rich-associated protein; Positive cofactor 2 glutamine/Q-rich-associated protein; positive cofactor 2, glutamine/Q-rich-associated protein; TIG-1; TPA inducible gene-1; TPA inducible protein; TPA-inducible gene 1 protein; trinucleotide repeat containing 7; Trinucleotide repeat-containing gene 7 protein
Gene Aliases: ARC105; CAG7A; CTG7A; MED15; PCQAP; TIG-1; TIG1; TNRC7
UniProt ID: (Human) Q96RN5
Entrez Gene ID: (Human) 51586
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.