Novus Biologicals
Manufacturer Code:NB1002357FR
Catalog # NB1002357FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 2150 and the C-terminus (residue 2212) of human Mediator of RNA Polymerase II Transcription subunit 12 (NP_005111.1). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator-recruited cofactor 240 kDa component; ARC240; ARC240 CAG repeat protein 45 CAGH45trinucleotide repeat containing 11 (THR-associated protein 230kDa subunit) HOPAFGS1 KIAA0192FG syndrome 1 mediator complex subunit 12OPA-containing protein OKSsubunit 12 homolog thyroid hormone receptor-associated protein 230 kDa subunit Trap230 TRAP230mediator of RNA polymerase II transcription subunit 12 homolog (S. cerevisiae) trinucleotide repeat containing 11 (THR-associated protein 230 kDa subunit) Trinucleotide repeat-containing gene 11 protein; CAG repeat protein 45; human opposite paired; Mediator complex subunit 12; Mediator of RNA polymerase II transcription subunit 12; mediator of RNA polymerase II transcription, subunit 12 homolog; OPA-containing protein; putative mediator subunit 12; Thyroid hormone receptor-associated protein complex 230 kDa component; thyroid hormone receptor-associated protein, 230 kDa subunit; Trap230; trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit); Trinucleotide repeat-containing gene 11 protein
Gene Aliases: ARC240; CAGH45; FGS1; HOPA; KIAA0192; MED12; MED12S; OHDOX; OKS; OPA1; TNRC11; TRAP230
UniProt ID: (Human) Q93074
Entrez Gene ID: (Human) 9968
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.