Abnova
Manufacturer Code:PAB28711
Catalog # 89-130-228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MED12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MED12.
For Research Use Only
Protein Aliases: Activator-recruited cofactor 240 kDa component; ARC240; CAG repeat protein 45; CAGH45; FGS1; HOPA; human opposite paired; KIAA0192; Mediator complex subunit 12; Mediator of RNA polymerase II transcription subunit 12; mediator of RNA polymerase II transcription, subunit 12 homolog; OKS; OPA-containing protein; OPA1; putative mediator subunit 12; Thyroid hormone receptor-associated protein complex 230 kDa component; thyroid hormone receptor-associated protein, 230 kDa subunit; TNRC11; TRAP230; trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit); Trinucleotide repeat-containing gene 11 protein
Gene Aliases: ARC240; CAGH45; FGS1; HOPA; KIAA0192; MED12; MED12S; OHDOX; OKS; OPA1; TNRC11; TRAP230
UniProt ID: (Human) Q93074
Entrez Gene ID: (Human) 9968
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.