Invitrogen
Manufacturer Code:PA535243
Catalog # PIPA535243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 201-229 amino acids from the central region of human MECR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-enoyl thioester reductase; Enoyl-[acyl-carrier-protein] reductase, mitochondrial; homolog of yeast 2-enoyl thioester reductase; HsNrbf-1; MECRNBRF1 Trans-2-enoyl-CoA reductase mitochondrial Trans-2-enoyl-CoA reductase mitochondrial Nuclear receptor-binding factor 1; mitochondrial 2-enoyl thioester reductase; nuclear receptor binding factor 1; Nuclear receptor-binding factor 1
Gene Aliases: CGI-63; FASN2B; MECR; NBRF1; NRBF1
UniProt ID: (Human) Q9BV79
Entrez Gene ID: (Human) 51102
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.