Proteintech
Manufacturer Code:149321AP150UL
Catalog # 50-172-9847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MECR Fusion Protein Ag6738 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-enoyl thioester reductase; CGI 63 FASN2B HsNrbf 1 MECR NBRF1 NRBF 1 NRBF1; Enoyl-[acyl-carrier-protein] reductase, mitochondrial; homolog of yeast 2-enoyl thioester reductase; HsNrbf-1; mitochondrial 2-enoyl thioester reductase; NRBF-1; nuclear receptor binding factor 1; Nuclear receptor-binding factor 1; trans-2-enoyl-CoA reductase, mitochondrial
Gene Aliases: AI195831; CGI-63; FASN2B; MECR; NBRF1; NRBF1
UniProt ID: (Human) Q9BV79, (Mouse) Q9DCS3, (Rat) Q9Z311
Entrez Gene ID: (Human) 51102, (Mouse) 26922, (Rat) 29470
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.