Proteintech
Manufacturer Code:108611AP150UL
Catalog # 50-172-6664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MECP2 Fusion Protein Ag1309 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AUTSX3 MeCp 2 protein MECP2 Methyl CpG binding protein 2 MRX16 MRX79 MRXS13 MRXSL PPMX RTS RTT; MeCp-2 protein; methyl CpG binding protein 2; Methyl-CpG-binding protein 2; testis tissue sperm-binding protein Li 41a
Gene Aliases: 1500041B07Rik; AUTSX3; D630021H01Rik; Mbd5; MECP2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; WBP10
UniProt ID: (Human) P51608, (Mouse) Q9Z2D6, (Rat) Q00566
Entrez Gene ID: (Human) 4204, (Mouse) 17257, (Rat) 29386
Molecular Function:
methyltransferase
nuclease
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.