Invitrogen
Manufacturer Code:PA578624
Catalog # PIPA578624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human ME3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); Malic enzyme 3; malic enzyme 3, NADP(+)-dependent, mitochondrial; malic enzyme, NADP+-dependent, mitochondrial; mitochondrial NADP(+)-dependent malic enzyme 3; NADP-dependent malic enzyme, mitochondrial; NADP-ME; NADP-ME malic enzyme 3 NADP(+)-dependent mitochondrial; pyruvic-malic carboxylase
Gene Aliases: 1700020C08Rik; B230207H15Rik; ME3; NADP-ME
UniProt ID: (Human) Q16798, (Mouse) Q8BMF3
Entrez Gene ID: (Human) 10873, (Mouse) 109264
Molecular Function: acyltransferase decarboxylase dehydrogenase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.