Invitrogen
Manufacturer Code:PA5101699
Catalog # PIPA5101699
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ME3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700020C08Rik B230207H15Rik HME Macrophage elastase malate dehydrogenase malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)) Malic enzyme 3 malic enzyme 3 NADP(+)-dependent mitochondrial malic enzyme NADP+-dependent mitochondrial ME Me3 mitochondrial malic enzyme 3 mitochondrial NADP(+)-dependent malic enzyme 3 MME MMP3 NADP-dependent malic enzyme mitochondrial NADP-ME pyruvic-malic carboxylase; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); Malic enzyme 3; malic enzyme 3, NADP(+)-dependent, mitochondrial; malic enzyme, NADP+-dependent, mitochondrial; mitochondrial NADP(+)-dependent malic enzyme 3; NADP-dependent malic enzyme, mitochondrial; NADP-ME; pyruvic-malic carboxylase
Gene Aliases: 1700020C08Rik; B230207H15Rik; ME3; NADP-ME
UniProt ID: (Human) Q16798, (Mouse) Q8BMF3
Entrez Gene ID: (Human) 10873, (Mouse) 109264
Molecular Function:
acyltransferase
decarboxylase
dehydrogenase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.