Abnova
Manufacturer Code:PAB27010
Catalog # 89-128-659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human ME3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ME3.
For Research Use Only
Protein Aliases: FLJ34862; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); Malic enzyme 3; malic enzyme 3, NADP(+)-dependent, mitochondrial; malic enzyme, NADP+-dependent, mitochondrial; mitochondrial NADP(+)-dependent malic enzyme 3; NADP-dependent malic enzyme, mitochondrial; NADP-ME; pyruvic-malic carboxylase
Gene Aliases: ME3; NADP-ME
UniProt ID: (Human) Q16798
Entrez Gene ID: (Human) 10873
Molecular Function:
acyltransferase
decarboxylase
dehydrogenase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.