Invitrogen
Manufacturer Code:MA537745
Catalog # PIMA537745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 970CT2.1.1.2 |
Immunogen | KLH conjugated synthetic peptide between 522-556 amino acids from the C-terminal region of human ME2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AW120568 D030040L20Rik malate dehydrogenase malate dehydrogenase (oxaloacetate-decarboxylating) malic enzyme 2 malic enzyme 2 NAD(+)-dependent mitochondrial ME2 NAD-dependent malic enzyme mitochondrial NAD-ME ODS1 pyruvic-malic carboxylase; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating); Malic enzyme 2; malic enzyme 2, NAD(+)-dependent, mitochondrial; NAD-dependent malic enzyme, mitochondrial; NAD-ME; pyruvic-malic carboxylase
Gene Aliases: AW120568; D030040L20Rik; ME2; NAD-ME; ODS1
UniProt ID: (Human) P23368, (Mouse) Q99KE1
Entrez Gene ID: (Human) 4200, (Mouse) 107029
Molecular Function:
acyltransferase
decarboxylase
dehydrogenase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.