Novus Biologicals
Manufacturer Code:NBP15476520UL
Catalog # NBP15476520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ME2(malic enzyme 2 NAD(+)-dependent mitochondrial) The peptide sequence was selected from the N terminal of ME2. Peptide sequence MAFTLQERQMLGLQGLLPPKIETQDIQALRFHRNLKKMTSPLEKYIYIMG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.1.1 EC 1.1.1.38 malate dehydrogenase Malic enzyme 2 malic enzyme 2 NAD(+)-dependent mitochondrial NAD-dependent malic enzyme mitochondrial NAD-ME ODS1 pyruvic-malic carboxylase; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating); Malic enzyme 2; malic enzyme 2, NAD(+)-dependent, mitochondrial; NAD-dependent malic enzyme, mitochondrial; NAD-ME; pyruvic-malic carboxylase
Gene Aliases: ME2; ODS1
UniProt ID: (Human) P23368
Entrez Gene ID: (Human) 4200
Molecular Function:
acyltransferase
decarboxylase
dehydrogenase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.