Novus Biologicals
Manufacturer Code:NBP243669AF532
Catalog # NP243669A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 15611 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ME1. The exact sequence is proprietary. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 1.1.1 EC 1.1.1.40 HUMNDME malate dehydrogenase Malic enzyme 1 malic enzyme 1 NADP(&plus)-dependent cytosolic malic enzyme 1 soluble Malic enzyme cytoplasmic MES NADP-dependent malic enzyme NADP-ME pyruvic-malic carboxylase; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); Malic enzyme 1; malic enzyme 1, NADP(+)-dependent, cytosolic; malic enzyme 1, soluble; Malic enzyme, cytoplasmic; NADP-dependent malic enzyme; NADP-ME; pyruvic-malic carboxylase
Gene Aliases: HUMNDME; ME1; MES
UniProt ID: (Human) P48163
Entrez Gene ID: (Human) 4199
Molecular Function: acyltransferase decarboxylase dehydrogenase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.