Novus Biologicals
Manufacturer Code:NBP303234100UL
Catalog # NB093586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human MDR3/ABCB4 (NP_061337.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC21 ATP-binding cassette sub-family B member 4 ATP-binding cassette sub-family B (MDR/TAP) member 4 EC 3.6.3 EC 3.6.3.44 MDR2 MDR3MDR2/3 multidrug resistance protein 3 multiple drug resistance 3 P glycoprotein 3/multiple drug resistance 3 PFIC-3 P-glycoprotein 3 P-glycoprotein-3/multiple drug resistance-3 PGY3GBD1; ATP-binding cassette sub-family B member 4; ATP-binding cassette, sub-family B (MDR/TAP), member 4; Multidrug resistance protein 3; multiple drug resistance 3; P glycoprotein 3/multiple drug resistance 3; P-glycoprotein 3; P-glycoprotein-3/multiple drug resistance-3; Phosphatidylcholine translocator ABCB4
Gene Aliases: ABC21; ABCB4; GBD1; ICP3; MDR2; MDR2/3; MDR3; PFIC-3; PGY3
UniProt ID: (Human) P21439
Entrez Gene ID: (Human) 5244
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.