Novus Biologicals
Manufacturer Code:NBP272605FR
Catalog # NBP272605FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G5 |
Immunogen | Full length recombinant protein of human MDMX (NP_002384) produced in HEK293T cells. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp781B1423 double minute 4 homolog Double minute 4 protein double minute 4 human homolog of p53-binding protein HDMX Mdm2-like p53-binding protein Mdm4 p53 binding protein homolog (mouse) Mdm4 transformed 3T3 cell double minute 4 p53 binding protein Mdm4 transformed 3T3 cell double minute 4 p53 binding protein (mouse) MDM4-related protein 1 MDMXMGC132766 mouse double minute 4 human homolog of p53-binding protein MRP1 p53-binding protein Mdm4 protein Mdm4 Protein Mdmx; Double minute 4 protein; double minute 4, human homolog of; p53-binding protein; Mdm2-like p53-binding protein; Mdm4 p53 binding protein homolog; MDM4 protein variant G; MDM4 protein variant Y; MDM4-related protein 1; p53-binding protein Mdm4; Protein Mdm4; Protein Mdmx
Gene Aliases: HDMX; MDM4; MDMX; MRP1
UniProt ID: (Human) O15151
Entrez Gene ID: (Human) 4194
Molecular Function: DNA binding protein chromatin/chromatin-binding protein ligase nucleic acid binding ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.