Novus Biologicals
Manufacturer Code:NBP297044AF405
Catalog # NB041740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant MDMX (NP_002384.2 Met1-Asp134). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781B1423 double minute 4 homolog Double minute 4 protein double minute 4 human homolog of p53-binding protein HDMX Mdm2-like p53-binding protein Mdm4 p53 binding protein homolog (mouse) Mdm4 transformed 3T3 cell double minute 4 p53 binding protein Mdm4 transformed 3T3 cell double minute 4 p53 binding protein (mouse) MDM4-related protein 1 MDMXMGC132766 mouse double minute 4 human homolog of p53-binding protein MRP1 p53-binding protein Mdm4 protein Mdm4 Protein Mdmx; Double minute 4 protein; double minute 4, human homolog of; p53-binding protein; Mdm2-like p53-binding protein; Mdm4 p53 binding protein homolog; MDM4 protein variant G; MDM4 protein variant Y; MDM4-related protein 1; p53-binding protein Mdm4; Protein Mdm4; Protein Mdmx
Gene Aliases: HDMX; MDM4; MDMX; MRP1
UniProt ID: (Human) O15151
Entrez Gene ID: (Human) 4194
Molecular Function: DNA binding protein chromatin/chromatin-binding protein ligase nucleic acid binding ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.