Proteintech
Manufacturer Code:179141AP150UL
Catalog # 50-173-1872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MDMX Fusion Protein Ag12319 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: double minute 4 homolog; Double minute 4 protein; Double minute 4 protein HDMX Mdm2 like p53 binding protein MDM4 MDMX MRP1 p53 binding protein Mdm4 Protein Mdm4 Protein Mdmx; double minute 4, human homolog of; p53-binding protein; Mdm2-like p53-binding protein; Mdm4 p53 binding protein homolog; MDM4 protein variant G; MDM4 protein variant Y; MDM4-related protein 1; p53-binding protein Mdm4; Protein Mdm4; Protein Mdmx; transformed mouse 3T3 cell double minute 4
Gene Aliases: 4933417N07Rik; AA414968; AL023055; AU018793; AU021806; C85810; HDMX; MDM4; MDMX; MRP1
UniProt ID: (Human) O15151, (Rat) Q5XIN1, (Mouse) O35618
Entrez Gene ID: (Human) 4194, (Rat) 304798, (Mouse) 17248
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
ligase
nucleic acid binding
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.