OriGene
Manufacturer Code:CF505736
Catalog # 50-166-3879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D11 |
Immunogen | Full length human recombit protein of human MDM4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Double minute 4 protein; double minute 4, human homolog of; p53-binding protein; HDMX MDMX MRP1; Mdm2-like p53-binding protein; Mdm4 p53 binding protein homolog; MDM4 protein variant G; MDM4 protein variant Y; MDM4-related protein 1; p53-binding protein Mdm4; Protein Mdm4; Protein Mdmx
Gene Aliases: HDMX; MDM4; MDMX; MRP1
UniProt ID: (Human) O15151
Entrez Gene ID: (Human) 4194
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
ligase
nucleic acid binding
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.