Proteintech
Manufacturer Code:161711AP150UL
Catalog # 50-173-0769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: double minute 1; Mdm1 nuclear protein homolog; MDM1 Nuclear protein MDM1; Mdm4 transformed 3T3 cell double minute 1 protein; Mdm4, transformed 3T3 cell double minute 1, p53 binding protein; Mouse double minute 1; nuclear protein double minute 1; Nuclear protein MDM1; transformed mouse 3T3 cell double minute 1
Gene Aliases: Arrd2; Mdm-1; MDM1
UniProt ID: (Human) Q8TC05, (Rat) Q5PQN4, (Mouse) Q9D067
Entrez Gene ID: (Human) 56890, (Rat) 314859, (Mouse) 17245
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.