Abnova
Manufacturer Code:H00056890K
Catalog # 89-209-197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MDM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MDM1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ95264; Mdm1 nuclear protein homolog; Mdm4, transformed 3T3 cell double minute 1, p53 binding protein; nuclear protein double minute 1; Nuclear protein MDM1
Gene Aliases: MDM1
UniProt ID: (Human) Q8TC05
Entrez Gene ID: (Human) 56890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.