Invitrogen
Manufacturer Code:PA589278
Catalog # PIPA589278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 59-338 of human MDH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: malate dehydrogenase 2, NAD (mitochondrial); Malate dehydrogenase, mitochondrial; mitochondrial malate dehydrogenase 2, NAD; testicular tissue protein Li 120
Gene Aliases: M-MDH; MDH; Mdh-2; MDH2; MGC:3559; Mor-1; MOR1
UniProt ID: (Human) P40926, (Mouse) P08249
Entrez Gene ID: (Human) 4191, (Mouse) 17448
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.