Proteintech
Manufacturer Code:665051IG150UL
Catalog # 50-173-6927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F9A2 |
Immunogen | MDH1 Fusion Protein Ag8744 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytosolic malate dehydrogenase; Cytosolic malate dehydrogenase MDH s MDH1 MDHA MOR2; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; malate dehydrogenase 1, NAD (soluble); Malate dehydrogenase, cytoplasmic; malate dehydrogenase, soluble; soluble malate dehydrogenase
Gene Aliases: B230377B03Rik; D17921; HEL-S-32; MDH-s; MDH1; MDHA; MGC:1375; Mor-2; MOR2
UniProt ID: (Human) P40925, (Mouse) P14152
Entrez Gene ID: (Human) 4190, (Mouse) 17449
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.