Invitrogen
Manufacturer Code:PA5100104
Catalog # PIPA5100104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MDH1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B230377B03Rik Cytosolic malate dehydrogenase D17921 Diiodophenylpyruvate reductase epididymis secretory protein Li 32 HEL-S-32 malate dehydrogenase (EC 1.1.1.37) malate dehydrogenase 1 malate dehydrogenase 1 NAD (soluble) malate dehydrogenase cytoplasmic malate dehydrogenase cytoplasmic malate dehydrogenase peroxisomal malate dehydrogenase soluble Malate dehydrogenase-like enzyme Mdh Mdh1 MDHA Mdhl MDH-s MDL1 MGC:1375 Mor2 Mor-2 soluble malate dehydrogenase; Cytosolic malate dehydrogenase; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; malate dehydrogenase 1, NAD (soluble); Malate dehydrogenase, cytoplasmic; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; soluble malate dehydrogenase
Gene Aliases: B230377B03Rik; D17921; HEL-S-32; Mdh; MDH-s; MDH1; MDHA; Mdhl; MDL1; MGC:1375; Mor-2; MOR2
UniProt ID: (Human) P40925, (Mouse) P14152, (Rat) O88989
Entrez Gene ID: (Human) 4190, (Mouse) 17449, (Rat) 24551
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.