Novus Biologicals
Manufacturer Code:NBP272604AF350
Catalog # NP272604A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8C10 |
Immunogen | Full length recombinant protein of human MDA5 (NP_071451) produced in HEK293T cells. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CADM-140 autoantigen; CADM-140 autoantigen Clinically amyopathic dermatomyositis autoantigen 140 kDa DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide EC 3.6.4.13 Helicard Helicase with 2 CARD domains Hlcd IDDM19 interferon induced with helicase C domain 1 interferon-induced helicase C domain-containing protein 1 Interferon-induced with helicase C domain protein 1 MDA-5MGC133047 MDA5RNA helicase-DEAD box protein 116 melanoma differentiation associated protein-5 Melanoma differentiation-associated protein 5 Murabutide down-regulated protein RH116; Clinically amyopathic dermatomyositis autoantigen 140 kDa; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; Helicard; Helicase with 2 CARD domains; interferon induced, with helicase C domain 1; Interferon-induced helicase C domain-containing protein 1; Interferon-induced with helicase C domain protein 1; MDA-5; melanoma differentiation associated protein-5; melanoma differentiation-associated gene 5; Melanoma differentiation-associated protein 5; Murabutide down-regulated protein; RIG-I-like receptor 2; RLR-2; RNA helicase-DEAD box protein 116
Gene Aliases: AGS7; Hlcd; IDDM19; IFIH1; MDA-5; MDA5; RH116; RLR-2; SGMRT1
UniProt ID: (Human) Q9BYX4
Entrez Gene ID: (Human) 64135
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.