Invitrogen
Manufacturer Code:MA536112
Catalog # PIMA536112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A2E11 |
Immunogen | Recombinant protein within human MCU aa 50-320. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2010012O16Rik AV064928 C10orf42 Calcium uniporter protein mitochondrial CCDC109A coiled-coil domain containing 109A coiled-coil domain-containing protein 109A D130073L02Rik Gm64 HsMCU MCU mitochondrial calcium uniporter; Calcium uniporter protein, mitochondrial; Coiled-coil domain-containing protein 109A; HsMCU
Gene Aliases: C10orf42; CCDC109A; MCU
UniProt ID: (Human) Q8NE86
Entrez Gene ID: (Human) 90550
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.