Invitrogen
Manufacturer Code:PA568153
Catalog # PIPA568153
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human MCU. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C10orf42 CCDC109A; Calcium uniporter protein, mitochondrial; coiled-coil domain containing 109A; Coiled-coil domain-containing protein 109A; HsMCU
Gene Aliases: 2010012O16Rik; AV064928; C10orf42; CCDC109A; D130073L02Rik; Gm64; MCU
UniProt ID: (Human) Q8NE86, (Mouse) Q3UMR5
Entrez Gene ID: (Human) 90550, (Mouse) 215999
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.