Invitrogen
Manufacturer Code:PA531020
Catalog # PIPA531020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 181 of Human MCT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: malignant T cell amplified sequence 1; malignant T cell-amplified sequence 1; malignant T-cell amplified sequence 1; Malignant T-cell-amplified sequence 1; MCT-1; MCT-1 MCT1 maligt T cell-amplified sequence 1 multiple copies T-cell maligcies; multiple copies in T-cell lymphoma-1; Multiple copies T-cell malignancies; Multiple copies T-cell malignancies 1
Gene Aliases: 1500019M23Rik; MCT-1; MCT1; MCTS1
UniProt ID: (Human) Q9ULC4, (Rat) Q4G009, (Mouse) Q9DB27
Entrez Gene ID: (Human) 28985, (Rat) 302500, (Mouse) 68995
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.