Novus Biologicals
Manufacturer Code:NBP2852650.1ML
Catalog # NB033194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human MCT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCT 2; MCT 2 MCT2solute carrier family 16 (monocarboxylic acid transporters) member 7 monocarboxylate transporter 2 Solute carrier family 16 member 7 solute carrier family 16 member 7 (monocarboxylic acid transporter 2); Monocarboxylate transporter 2; solute carrier family 16 (monocarboxylate transporter), member 7; Solute carrier family 16 member 7; solute carrier family 16, member 7 (monocarboxylic acid transporter 2)
Gene Aliases: MCT2; SLC16A7
UniProt ID: (Human) O60669
Entrez Gene ID: (Human) 9194
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.