Invitrogen
Manufacturer Code:PA576687
Catalog # PIPA576687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human MCT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ36745HHF7MCT 1MCTMCTMGC44475Monocarboxylate transporter 1Monocarboxylate transporterMonocarboxylate transporter isoform 1Monocarboxylic acid transporter 1MOT1&lowbarHUMANSlc16a1SLC16A1 proteinSolute carrier family 16 (monocarboxylic acid transporters) member 1Solute carrier family 16 (monocarboxylic acid transporters) member 1Solute carrier family 16 member 1 (monocarboxylic acid transporter 1)Solute carrier family 16 member 1; MCT 1; Monocarboxylate transporter 1; solute carrier family 16 (monocarboxylate transporter), member 1; solute carrier family 16 (monocarboxylic acid transporters), member 1; Solute carrier family 16 member 1; solute carrier family 16, member 1 (monocarboxylic acid transporter 1)
Gene Aliases: AL022710; HHF7; MCT; MCT1; MCT1D; SLC16A1
UniProt ID: (Human) Q9NSJ9, (Mouse) P53986
Entrez Gene ID: (Human) 6566, (Mouse) 20501
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.