Invitrogen
Manufacturer Code:PA512335
Catalog # PIPA512335
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 430-460 amino acids from the C-terminal region of human MCT1 (SLC16A1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCT 1; Monocarboxylate transporter 1; Monocarboxylate transporter 1 MCT 1 Solute carrier family 16 member 1 SLC16A1 MCT1; solute carrier family 16 (monocarboxylate transporter), member 1; solute carrier family 16 (monocarboxylic acid transporters), member 1; Solute carrier family 16 member 1; solute carrier family 16, member 1 (monocarboxylic acid transporter 1)
Gene Aliases: HHF7; MCT; MCT1; MCT1D; SLC16A1
UniProt ID: (Human) Q9NSJ9
Entrez Gene ID: (Human) 6566
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.