Invitrogen
Manufacturer Code:PA558639
Catalog # PIPA558639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MCRS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 58 kDa microspherule protein; cell cycle-regulated factor (78 kDa); Cell cycle-regulated factor p78; INO80 complex subunit J; INO80 complex subunit Q; MCRS2; Microspherule protein 1; Microspherule protein 1 58 kDa microspherule protein Cell cycle-regulated factor p78 INO80 complex subunit J MCRS2 MCRS1 INO80Q MSP58; nucleolar protein
Gene Aliases: C78274; ICP22BP; INO80Q; MCRS1; MCRS2; MSP58; P78
UniProt ID: (Human) Q96EZ8, (Mouse) Q99L90
Entrez Gene ID: (Human) 10445, (Rat) 300222, (Mouse) 51812
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.