Invitrogen
Manufacturer Code:M810
Catalog # PIM810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8C12 |
Immunogen | Recombit CCL13 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 13; C-C motif chemokine 13, long chain; C-C motif chemokine 13, medium chain; C-C motif chemokine 13, short chain; chemokine (C-C motif) ligand 13; Chemokine (C-C Motif) Ligand 13 Small inducible cytokine subfamily A (Cys-Cys) member 13 C-C Motif Chemokine 13 New CC Chemokine 1 CK-Beta-10 CKb10 Monocyte Chemoattractant Protein 4 Monocyte chemotatic protein 4 MCP4 MCP-4 NCC1 NCC-1 SCYA13 SCYL1; CK-beta-10; MCP-4; Monocyte chemoattractant protein 4; Monocyte chemotactic protein 4; NCC-1; new CC chemokine 1; small inducible cytokine subfamily A (Cys-Cys), member 13; Small-inducible cytokine A13
Gene Aliases: CCL13; CKb10; MCP-4; MCP4; NCC-1; NCC1; SCYA13; SCYL1
UniProt ID: (Human) Q99616
Entrez Gene ID: (Human) 6357
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.