Invitrogen
Manufacturer Code:PA578932
Catalog # PIPA578932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human MCP4 recombit protein (Position: Q24-T98). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 13; C-C motif chemokine 13 CK-beta-10 Monocyte chemoattractant protein 4 Monocyte chemotactic protein 4 MCP-4 NCC-1 Small-inducible cytokine A13 C-C motif chemokine 13 long chain C-C motif chemokine 13 medium chain C-C motif chemokine 13 short chain CCL13 MCP4 NCC1 SCYA13; C-C motif chemokine 13, long chain; C-C motif chemokine 13, medium chain; C-C motif chemokine 13, short chain; chemokine (C-C motif) ligand 13; CK-beta-10; MCP-4; Monocyte chemoattractant protein 4; Monocyte chemotactic protein 4; NCC-1; new CC chemokine 1; small inducible cytokine subfamily A (Cys-Cys), member 13; Small-inducible cytokine A13
Gene Aliases: CCL13; CKb10; MCP-4; MCP4; NCC-1; NCC1; SCYA13; SCYL1
UniProt ID: (Human) Q99616
Entrez Gene ID: (Human) 6357
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.