Invitrogen
Manufacturer Code:MA530229
Catalog # PIMA530229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 63 |
Immunogen | Recombit Human MCP-3/CCL7 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 7; chemokine (C-C motif) ligand 7; FIC MARC MCP-3 MCP3 MGC138463 MGC138465 NC28 SCYA6 SCYA7; MCP-3; Monocyte chemoattractant protein 3; Monocyte chemotactic protein 3; NC28; small inducible cytokine A7 (monocyte chemotactic protein 3); Small-inducible cytokine A7
Gene Aliases: CCL7; FIC; MARC; MCP-3; MCP3; NC28; SCYA6; SCYA7
UniProt ID: (Human) P80098
Entrez Gene ID: (Human) 6354
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.