Invitrogen
Manufacturer Code:PA586885
Catalog # PIPA586885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in Human CCL7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 7; C-C motif chemokine 7 Monocyte chemoattractant protein 3 Monocyte chemotactic protein 3 MCP-3 NC28 Small-inducible cytokine A7 CCL7 MCP3 SCYA6 SCYA7; chemokine (C-C motif) ligand 7; chemotactic protein-3; Intercrine/chemokine MARC; MCP-3; Monocyte chemoattractant protein 3; Monocyte chemotactic protein 3; NC28; Protein FIC; RANTES/sis homolog; small inducible cytokine A7; small inducible cytokine A7 (monocyte chemotactic protein 3); Small-inducible cytokine A7
Gene Aliases: CCL7; FIC; MARC; MCP-3; MCP3; NC28; SCYA6; SCYA7
UniProt ID: (Human) P80098, (Rat) Q9QXY8, (Mouse) Q03366
Entrez Gene ID: (Human) 6354, (Rat) 287561, (Mouse) 20306
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.