Invitrogen
Manufacturer Code:PA546952
Catalog # PIPA546952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse CCL7/MCP-3/MARC Gln24-Pro97 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 7; FIC MARC MGC138463 MGC138465 NC28 SCYA6 SCYA7; Intercrine/chemokine MARC; MCP-3; Monocyte chemoattractant protein 3; Monocyte chemotactic protein 3; Protein FIC; RANTES/sis homolog; small inducible cytokine A7; Small-inducible cytokine A7
Gene Aliases: Ccl7; Fic; marc; MCP-3; Mcp3; Scya7
UniProt ID: (Mouse) Q03366
Entrez Gene ID: (Mouse) 20306
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.