Invitrogen
Manufacturer Code:701174
Catalog # PI701174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 3H18L3 |
Immunogen | proprietary |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 8; C-C motif chemokine 8 SCYA10 SCYA8 HC14 Monocyte chemoattractant protein 2 Monocyte chemotactic protein 2 Small-inducible cytokine A8; chemokine (C-C motif) ligand 8; HC14; MCP-2; MCP-2(6-76); Monocyte chemoattractant protein 2; Monocyte chemotactic protein 2; small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2); Small-inducible cytokine A8
Gene Aliases: CCL8; HC14; MCP-2; MCP2; SCYA10; SCYA8
UniProt ID: (Human) P80075
Entrez Gene ID: (Human) 6355
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.