Invitrogen
Manufacturer Code:MA124663
Catalog # PIMA124663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 35509.11 |
Immunogen | Recombit human monocyte chemotactic protein 2 (MCP2) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 8; chemokine (C-C motif) ligand 8; HC14; HC14 MCP-2 MCP2 SCYA10 SCYA8; MCP-2; MCP-2(6-76); Monocyte chemoattractant protein 2; Monocyte chemotactic protein 2; small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2); Small-inducible cytokine A8
Gene Aliases: CCL8; HC14; MCP-2; MCP2; SCYA10; SCYA8
UniProt ID: (Human) P80075
Entrez Gene ID: (Human) 6355
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.