Invitrogen
Manufacturer Code:M2MCP1I
Catalog # PIM2MCP1I
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | E10052 |
Immunogen | Recombit human MCP-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 2; C-C motif chemokine 2 monocyte chemoattractant protein 1 monocyte chemoattractant protein-1 monocyte chemotactic and activating factor monocyte chemotactic protein 1 monocyte secretory protein JE small inducible cytokine A2 (monocyte chemotactic protein 1 homologous to mouse Sig-je) small inducible cytokine subfamily A (Cys-Cys) member 2 small-inducible cytokine A2; chemokine (C-C motif) ligand 2; HC11; MCAF; MCP-1; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; Monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; Monocyte secretory protein JE; Platelet-derived growth factor-inducible protein JE; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; Small-inducible cytokine A2
Gene Aliases: AI323594; CCL2; GDCF-2; HC11; HSMCR30; JE; MCAF; MCP-1; MCP1; SCYA2; Sigje; SMC-CF
UniProt ID: (Human) P13500, (Mouse) P10148
Entrez Gene ID: (Human) 6347, (Mouse) 20296
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.