Invitrogen
Manufacturer Code:PA568750
Catalog # PIPA568750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human MCM4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC21 CDC54 P1-CDC21 hCdc21 CDC21 homolog DNA replication licensing factor MCM4 MCM4 minichromosome maintece deficient 4 homolog of S. pombe cell devision cycle 21 minichromosome maintece deficient 4; CDC21 homolog; DNA replication licensing factor MCM4; homolog of S. pombe cell devision cycle 21; MCM4 minichromosome maintenance deficient 4; MCM4 minichromosome maintenance deficient 4, mitotin; mini chromosome maintenance deficient 4 homolog; minichromosome maintenance deficient 4; minichromosome maintenance deficient 4 homolog; P1-CDC21
Gene Aliases: 19G; AI325074; AU045576; cb1025; CDC21; CDC54; fc12c09; fj85g09; hCdc21; hm:zeh1616; mcdc21; MCM4; Mcmd4; mKIAA4003; NKCD; NKGCD; P1-CDC21; wu:fc12c09; wu:fj85g09; zeh1616
UniProt ID: (Human) P33991, (Mouse) P49717
Entrez Gene ID: (Rabbit) 100349822, (Dog) 477871, (Bovine) 504367, (Horse) 100051303, (Pig) 100156398, (Human) 4173, (Rat) 29728, (Guinea pig) 100724549, (Mouse) 17217, (Zebrafish) 337598
Molecular Function:
DNA binding protein
DNA helicase
helicase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.