Novus Biologicals
Manufacturer Code:NBP185728
Catalog # NBP185728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LPHTLLSRFDLIFLMLDPQDEAYDRRLAHHLVALYYQSEEQAEEELLDMAVLKDYIAYAHSTIMPRLSEEASQALIEAYVDMRKIGSSRGMVSAYPRQLESLIRLAEAHAKVRLSNKVEAIDVEEAKRLHREALKQSAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC21 CDC21 homolog CDC54 DNA replication licensing factor MCM4 EC 3.6.4.12 hCdc21 homolog of S. pombe cell devision cycle 21 MCM4 minichromosome maintenance deficient 4 MCM4 minichromosome maintenance deficient 4 (S. cerevisiae) MGC33310 minichromosome maintenance complex component 4 minichromosome maintenance deficient 4 P1-Cdc21; CDC21 homolog; DNA replication licensing factor MCM4; homolog of S. pombe cell devision cycle 21; minichromosome maintenance deficient 4; P1-CDC21
Gene Aliases: CDC21; CDC54; hCdc21; MCM4; NKCD; NKGCD; P1-CDC21
UniProt ID: (Human) P33991
Entrez Gene ID: (Human) 4173
Molecular Function: DNA binding protein DNA helicase helicase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.