Invitrogen
Manufacturer Code:14670137
Catalog # 14-670-137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Ab22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AW556805 BCL2 family apoptosis regulator BCL2L3 bcl2-L-3 Bcl-2-like protein 3 Bcl-2-related protein EAT/mcl1 EAT Induced myeloid leukemia cell differentiation protein Mcl-1 induced myeloid leukemia cell differentiation protein Mcl-1 homolog MCL1 Mcl-1 mcl1/EAT MCL1-ES MCL1L MCL1S MCL-1S MGC104264 MGC1839 myeloid cell leukemia 1 myeloid cell leukemia ES myeloid cell leukemia sequence 1 myeloid cell leukemia sequence 1 (BCL2-related) TM; Bcl-2-like protein 3; Bcl-2-related protein EAT/mcl1; Bcl2-L-3; Induced myeloid leukemia cell differentiation protein Mcl-1; mcl1/EAT; myeloid cell leukemia 1; myeloid cell leukemia ES; myeloid cell leukemia sequence 1 (BCL2-related)
Gene Aliases: bcl2-L-3; BCL2L3; EAT; Mcl-1; MCL1; MCL1-ES; mcl1/EAT; MCL1L; MCL1S; TM
UniProt ID: (Human) Q07820
Entrez Gene ID: (Human) 4170
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.