Invitrogen
Manufacturer Code:PA577492
Catalog # PIPA577492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CMDLQTSLLSTGPNASN corresponding to amino acid residues 7-20 of rat Melanin-concentrating hormone receptor 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor 24; G protein-coupled receptor 24; G-protein coupled receptor 24; Gpr24-9 protein coupled receptor; MCH receptor 1; MCH-1R; MCH-R1; MCH1 MCH1R GPCR24 SLC-1; MCH1R; MCHR; MCHR-1; Melanin-concentrating hormone receptor 1; SLC-1; Somatostatin receptor-like protein
Gene Aliases: AW049955; GPR24; MCH-1R; MCH1R; MCHR1; SLC-1; SLC1
UniProt ID: (Human) Q99705, (Mouse) Q8JZL2, (Rat) P97639
Entrez Gene ID: (Human) 2847, (Mouse) 207911, (Rat) 83567
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.