Novus Biologicals
Manufacturer Code:NBP1458550.025MG
Catalog # NB018591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-expressed rat SLC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 24; G protein-coupled receptor 24 GPR24SLC-1 G-protein coupled receptor 24 MCH receptor 1 MCH-1R MCH1RMCHR-1 MCH-R1 melanin-concentrating hormone receptor 1 MGC32129 SLC1MCHR Somatostatin receptor-like protein; G-protein coupled receptor 24; MCH receptor 1; MCH-1R; MCH1R; Melanin-concentrating hormone receptor 1; SLC-1; Somatostatin receptor-like protein
Gene Aliases: GPR24; MCH-1R; MCH1R; MCHR1; SLC-1; SLC1
UniProt ID: (Human) Q99705
Entrez Gene ID: (Human) 2847
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.