Abnova
Manufacturer Code:PAB16225
Catalog # 89-118-111
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human MCHR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of MCHR1.
For Research Use Only
Protein Aliases: G protein-coupled receptor 24; G-protein coupled receptor 24; GPR24; MCH receptor 1; MCH-1R; MCH1R; Melanin-concentrating hormone receptor 1; MGC32129; SLC-1; SLC1; Somatostatin receptor-like protein
Gene Aliases: GPR24; MCH-1R; MCH1R; MCHR1; SLC-1; SLC1
UniProt ID: (Human) Q99705
Entrez Gene ID: (Human) 2847
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.