Proteintech
Manufacturer Code:164991AP150UL
Catalog # 50-173-0990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MCEE Fusion Protein Ag9665 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DL methylmalonyl CoA racemase GLOD2 MCEE methylmalonyl CoA epimerase; DL-methylmalonyl-CoA racemase; glyoxalase domain containing 2; methylmalonyl CoA epimerase; Methylmalonyl-CoA epimerase, mitochondrial
Gene Aliases: 1110007A04Rik; GLOD2; MCEE
UniProt ID: (Human) Q96PE7, (Mouse) Q9D1I5
Entrez Gene ID: (Human) 84693, (Mouse) 73724, (Rat) 293829
Molecular Function: DNA binding protein DNA glycosylase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.