Invitrogen
Manufacturer Code:PA5106295
Catalog # PIPA5106295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MLYCD corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI324784 malonyl coenzyme A decarboxylase malonyl-CoA decarboxylase malonyl-CoA decarboxylase mitochondrial MCD MLYCD; malonyl coenzyme A decarboxylase; Malonyl-CoA decarboxylase, mitochondrial; MCD
Gene Aliases: AI324784; MCD; MLYCD
UniProt ID: (Human) O95822, (Rat) Q920F5, (Mouse) Q99J39
Entrez Gene ID: (Human) 23417, (Rat) 85239, (Mouse) 56690
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.