Novus Biologicals
Manufacturer Code:NBP297284G
Catalog # NB101579
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MCCC2 fragment. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-methylcrotonyl-CoA carboxylase 2; 3-methylcrotonyl-CoA carboxylase non-biotin-containing subunit; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta MCCase subunit beta MCCBEC 6.4.1.4 methylcrotonoyl-CoA carboxylase 2 (beta) methylcrotonoyl-Coenzyme A carboxylase 2 (beta) mitochondrial non-biotin containing subunit of 3-methylcrotonyl-CoA carboxylase; biotin carboxylase; MCCase subunit beta; methylcrotonoyl-CoA carboxylase 2 (beta); Methylcrotonoyl-CoA carboxylase beta chain, mitochondrial; methylcrotonoyl-Coenzyme A carboxylase 2 (beta); non-biotin containing subunit of 3-methylcrotonyl-CoA carboxylase; testicular secretory protein Li 29
Gene Aliases: MCCB; MCCC2
UniProt ID: (Human) Q9HCC0
Entrez Gene ID: (Human) 64087
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.